Learn More
Adam Equipment Adam Equipment TBB 610S Triple Beam Mechanical Balance, 610g Capacity, 0.1g Readability
Supplier: Adam Equipment TBB 610S
With a larger overall body than others in this category, the TBB is the sturdiest choice today in triple beam balances. Quality construction with a strong metal housing and a stainless steel pan ensure that your balance will last in the long run. Determining density of small samples is easy with the innovative tripod legs and weigh-below hook that come standard. An integral security bracket, compatible with Kensington type security lock and cable, is built into the balance housing for maximum protection. A protractor, ruler, and conversion chart are included right on the housing.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.