Learn More
Thermo Scientific™ Target Polyspring Insert
Polyspring inserts for 9mm HPLC and GC autosampler vials are available in a variety of configurations with usable volumes from 160μL to 3.5mL
Supplier: Thermo Scientific™ C4010630TM
Description
- Pulled point inserts are an economical choice for noncritical applications
- Nominal O.D. of 6mm to fit Target wide-opening vials
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
300 μL | |
100 |