Learn More
Fisherfinest™ Taped Biopsy Cassettes for Leica IP C and Sakura Cassette Printers
Pre-stacked, quick-load cassettes designed with efficient MicrOflow II™ reagent exchange technology
This item is not returnable.
View return policy
Description
- Validated by Leica Applications Lab for efficient, error-free use with Leica IP C and Sakura cassette printers
- Taped, pre-stack delivery method provides quick and efficient loading of the cassette magazines
- One-piece cassettes with frangible hinge for easy lid removal
- 45-Degree print surface for optimal workflow
- 0.67mm Pore
- Available in 6 popular colors that are optimized for use with 2D barcode scanning
Patented MicrOflow II™ technology features lateral vent design for maximum fluid exchange and efficient tissue processing, as well as a unique design with angled interior for easy access to the specimen.
Specifications
45° | |
Green | |
Biopsy Cassette |