Learn More
Perkin Elmer US LLC Copper Torch Ignitor Tape for Optima 2x00/4x00/5x00/7x00 DV
Supplier: Perkin Elmer US LLC N0775297
This copper ignitor tape is also included in the Scott/Cross-Flow II Sample Introduction Kit for Optima 2x00/4x00/5x00/7x00 DV (N0770731), the Cyclonic/Concentric Sample Introduction Kit for Optima 2x00/4x00/5x00/7x00 DV (N0770730), and the Organics Kit for Optima 4300 V/5300 V/7300 V(N0770732).