Learn More
Euthanex Tamper-Proof Single CO2 Flow Meter with DISS (0.5 - 12 LPM)
Supplier: Euthanex 512T1DS
This CO2 Flow Meter is set for a single flow rate, ideal for facilities that are working with a single cage size. Tamper Proof Flow Meters take the guess work out, and prevent the user performing the procedure incorrectly. Flow rate range 0.5 to 12 LPM. Ideal for single cages varying from mice to guinea pig. Connects directly to point of use CO2 source with a standard male DISS 1080 fitting. We also offer male DISS fitting in 1/8"""" and 1/4"""" NPT. The DISS includes an output line with quick disconnect for easy connection to our lids or induction chambers.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.