Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Talc, Powder, USP, Spectrum™ Chemical

Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Talc, Powder, USP is used as a carrier for pharmaceuticals with a disinfectant, astringent, anti-itching or cooling effect. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
CAS Max % | 1 |
Quantity | 2.5 kg |
Alkalinity | 0.4 mL |
Grade | USP |
Ignition Residue | 0.07 |
Packaging | Poly Pail |
Acidity | 0.3 mL |