Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tagg Industries Biocare Lock Box
SDP

Durable, impact-resistant ABS plastic box for after-hours pickup and delivery

Supplier:  Tagg Industries 50114

Encompass

Catalog No. 19-085506


Add to Cart

Description

Description

  • Hangs from the top of any office door by a 12 in. (30cm) over-the-door metal strap
  • Comes with a set of two keys

Order Info

L x W x D: 14 x 11 x 7 in. (35.6 x 27.9 x 17.8cm).

Specifications

Specifications

Biocare Lockbox
7 in.
For after-hours pickup and delivery of samples
28 cm
14 in.
ABS
18 cm
11 in.
Two keys, metal strap
36 cm
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit