Learn More
Cole-Parmer Extech 461825 Combination Optical Tachometer/Stroboscope
Supplier: Cole-Parmer 8770006
This meter is both a tachometer and a stroboscope. Use to measure rpm and speed, or motion analysis. Stroboscope mode features three selectable modes. Display flips with the meter for easy viewing. Meter automatically stores last reading, minimum and maximum in memory.