Learn More
Cole-Parmer Extech 461995 Combination Laser/contact Tachometer
Supplier: Cole-Parmer 8770010
These tachometers feature both contact and optical measurement capabilities. Memory button holds last reading up to 5 minutes and recalls minimum and maximum readings. Display flips with the unit to help identify measuring mode. Model 87700-10 features a laser sighting for spotting targets up to 6 1/2 feet (1.9 m) away.