Learn More
Cole-Parmer Shimpo MT-200 Digital Laser Compact Pocket Hand Tachometer
Supplier: Cole-Parmer 9880720
These rugged tachometers fit perfectly in the left or right hand, and are designed for use with a gloved hand. Units provide fourteen different units of measure including rpm, inch/minute, feet/minute to total length measurements like feet and meters, and more. Additional memory features include last, minimum, maximum, and average of results. Durable units are ideal for the most challenging industrial environments, and the compact design is small enough to fit into your shirt pocket. Model 98807-20 noncontact tachometer is easy to use and incorporates a laser that is accurate up to 6 ft (1.8 m) from the target, ensuring a safe distance from rotating parts.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.