Learn More
Biocare Medical, LLC Tacha Automated Hematoxylin - 500 ml
Supplier: Biocare Medical, LLC NMHEMM
Biocare progressive hematoxylin is water-based and is specially formulated for immunohistochemistry. It can be used for both paraffin and frozen sections. Tacha Automated Hematoxylin stain nuclei a beautiful sky-blue and provides the end-user high contrast staining for DAB, AEC, Bajoran Purple and Fast Red procedures. Tacha hematoxylin contains an anti-oxidant, and in most cases does not need filtering. It can also be used on an automated staining system.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.