Learn More
Thermo Scientific™ T7 RNA Polymerase (20 U/μL) PROMO
T7 RNA Polymerase is a DNA-dependent RNA polymerase with strict specificity for its double-stranded promoter.
Supplier: Thermo Scientific™ EP0111
Description
It catalyzes the 5' => 3' synthesis of RNA on either single-stranded DNA or double-stranded DNA downstream from it promoter.
- Incorporates modified nucleotides (e.g., aminoallyl-, biotin-, fluorescein-, digoxigenin-labeled nucleotides)
Recommended for:
Synthesis of unlabeled and labeled RNA that can be used:for hybridization, in vitro RNA translation; as aRNA, siRNA, substrate in RNase protection assays, template for genomic DNA sequencing.
Note:
Consensus promoter sequences: TAATACGACTCACTATA G GGAGA. The position in bold (+1) indicates the first nucleotide incorporated into RNA during transcription. Only bases at this position through +3 are critical for transcription, and they must be a G and a purine base, respectively.
Specifications
T7 RNA Polymerase | |
5000 Units | |
20 U/μL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.