Learn More
Synthware™ T-Type High Vacuum Hollow Plug Stopcock
GREENER_CHOICESDP
Precision ground with corresponding numbers on the barrel and plug.
Supplier:  Synthware™ S721754
 View more versions of this product
Catalog No. 31-502-052
Add to Cart
Description
Description
Specifications
Specifications
T-Type High Vacuum Valve
10 mm
Organic Synthesis
4 mm
Borosilicate Glass
Glass
Product Suggestions
Product Suggestions