Learn More
UFP TECHNOLOGIES INC T-TUBE ELBOW 90DEG 0.5IN OD
Supplier: UFP TECHNOLOGIES INC 101197
UFP Technologies' elbow, 90 degree, non-jacketed T-Tubes are sold per piece. Long radius and butt-welded unless specified otherwise. Item 101197 has a line OD of 0.5 inches.