Learn More
Med Vet International I-Glo Fluorescein Opthalmic Test Strips, 100/boxPH-FUL-GLO
Supplier: Med Vet International JOR1191
A great tool for detection of corneal injuries or defects. Every eye that has signs of pain should be checked with fluorescein. Tear film will wash dye across cornea. The area of the cornea injury will glow fluorescent.