Learn More
Cytiva (Formerly Pall Lab) T-series Centramate™ Cassette, Omega PES membrane, 10 kDa molecular weight cut-off (MWCO), 0.1 m2 effective filtration area (EFA)
Supplier: Cytiva OS010T12
T-series Centramate™ Cassette, Omega PES membrane, 10 kDa molecular weight cut-off (MWCO), 0.1 m2 effective filtration area (EFA). Pall Laboratory products are designed for laboratory applications only. This product is not approved for use in medical, clinical, surgical or other patient applications