Learn More
Cole-Parmer Digi-Sense K30-1-304 Thermocouple Wire Type K 30-Gauge, Glass Braid Insulation, 100-ft Spool
Supplier:  Cole-Parmer 0854103
Encompass_Preferred
Wire comes in 30 gauge size, ideal for fabricating your own probes or extension cables (meets ANSI and ASTM standards). Choose from wire with fluorinated ethylene propylene resin (FEP/FEP) or fiberglass materials as well as in 100-ft or 1000-ft lengths for any application.
Catalog No. 50-196-1524