Learn More
Cole-Parmer Digi-Sense K24-5-502 Thermocouple Wire Type K 24-Gauge, Polyvinyl Insulation, 100-ft Spool
Supplier:  Cole-Parmer 0854109
Encompass_Preferred
Make your own extension cables. Wire comes in 30 gauge size, ideal for fabricating your own probes or extension cables (meets ANSI and ASTM standards).
Catalog No. 50-196-1527