Learn More
ProteinSimple iCE System Suitability Kit, 10 vials
Supplier: ProteinSimple 102093
FC Cartridge System Suitability Kit. This kit includes a hemoglobin stock solution and 10 vials of a standard solution (carrier ampholytes and pI markers in 0.35% methyl cellulose).