Learn More
Electron Microscopy Sciences Syracuse Watch Glass 65 mm(OD) x 50 mm(ID) x 10 mm(Deep)
Supplier: Electron Microscopy Sciences 7157001
A clear watch glass which measures 65 mm(OD)x50 mm(ID)x 10 mm(Deep). The glass is grooved and has a recessed bottom which allows for stacking and prevents scratching. It is ideal for staining and specimen preparation.