Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synthware™ Chromatography Flow Control Adapters
High vacuum valve with metering tip to adjust air pressure
Supplier: Synthware™ C1301
Description
- 044mm high vacuum valve with metering tip to adjust air pressure
- With either a 24/40 inner standard taper joint or a 35/20 ball spherical joint
Specifications
Adapter | |
Ball spherical joint: 35/20 | |
Borosilicate Glass | |
15 to 25 psi |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction