Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Justrite™ Swivel Caster Set for Gas Cylinder Hand Trucks
Justrite™ Swivel Caster Set for Gas Cylinder Hand Trucks Post is a replacement caster set for single and double cylinder Justrite™ hand trucks
$49.80 - $72.49
Catalog Number | Mfr. No. | Description | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Material | Price | Quantity & Availability | ||||
19-085-860
|
Justrite™
35376 |
3 in. wheels | Polypropylene Alloy |
Each for $49.80
|
|
||||
19-085-867
|
Justrite™
35390 |
5 in. wheels | HDPE |
Each for $72.49
|
|
||||
Description
Two wheels with ball bearing steel swivel yokes, on a 0.5-inch threaded post with flat washer, lock washer and nutCompliance
OSHA, CGA P1, NFPA55
Warranty and Services
1 YEAR
Specifications
Swivel Caster Set | |
Gas cylinders |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title