Learn More
Eppendorf™ Swing Bucket Rotors
Perfect for phase-separation applications such as phenol-chloroform extraction
Specifications
Type | Swinging Bucket Rotor |
---|---|
Capacity | 24 x 1.5/2 mL |
Max. RCF | 16,049 x g |
Max. Speed | 12,700 rpm |
Catalog Number | Mfr. No. | Capacity | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity | Includes | Price | Quantity & Availability | ||||
05-414-315
|
Eppendorf™
5409715003 |
24 x 1.5/2 mL | QuickLock rotor lid, wind shield |
Each of 1 for $2,279.00
|
N/A | ||||
Description
- Designed for use with Eppendorf Centrifuges
Specifications
Swinging Bucket Rotor | |
24 x 1.5/2 mL | |
16,049 x g | |
12,700 rpm |