Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Swift™ Advanced Stereo Zoom Microscope
SCI_ED

The Swift™ Optical Advanced Stereo Zoom Microscope offers convenience and versatility of viewing items at 10X magnification then zoom continuously to maximum detail at 40X magnification.  It is ideal for educational, laboratory, research and industrial use.

Supplier:  Swift™ M29TZSM99CL

Catalog No. S23868

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View alternate product


Includes: Accessories : Dustcover, rubber eyeshields, glass stage plate, black and white contrast stage plate

Description

Description

  • Viewing head rotates 360°
  • Zoom 1X-4X
  • Acromatic, color-corrected lenses
  • Tension adjustment eliminates focus drift
  • Working distance - 80mm
  • Large dual focusing knobs with slip clutch to prevent damage to mechanism
  • Top and bottom LED illumination
  • C-mount port for easy addition of optional video C-mount and SLR cameras
Specifications

Specifications

10X to 40X
M29TZ-SM99CL
LED
Microscope
Trinocular
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit