Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Swan™ Petroleum Jelly

Protects minor cuts, burns and chapped skin.

Supplier:  Medique 15221

Catalog No. 19-090-843



Edge
Add to Cart

Description

Description

  • Skin protectant
  • With white petrolatum, USP 100%
Specifications

Specifications

Skin Protectant
Protects minor cuts, burns and chapped skin
White Petrolatum USP 100%
13 oz.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit