Learn More
Julabo USA Inc Each, SW23 shaking water bath
Supplier: Julabo USA Inc 9550323.2
Bath/Circulators, Julabo, JULABO SW shaking water baths for unsupervised sample incubation. Stainless steel construction with an operating range from +20...+99.9 °C. User-friendly operation via keypad with bright LED display. Electronic timer from 1 min to 9h 59min. Integrated RS232 interface for integration to PC software for control and data logging. Early low water level warning system. Early warning system with high and low temperature limits ensures that samples maintain the proper temperature. A removable shaking carriage for easy sample removal. A wide range of accessories, including bath covers, are available. Shaking frequency adjustable from 20 to 200 rpm. Shaking stroke = 15 mm. Lid sold separately.SW22 unit with ±0.2 °C stability. SW23 incorporates a water circulation pump for a stability of ±0.02 °C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.