Learn More
Sheldon Manufacturing Inc SVAC-1E VAC OVEN 115V
Supplier: Sheldon Manufacturing Inc SVAC1E
ECONOMY VACUUM OVEN, 0.6 CU FT, 115V SHEL LAB vacuum ovens provide exceptional capabilities for fast and gentle drying of heat sensitive materials. They prevent residue accumulation on products and also allow safe drying of materials at lower temperatures than typically required when using convection ovens. Please note that this vacuum oven does not include a pump or related accessories. Contact your SHEL LAB distributor for assistance with choosing the ideal pump and accessories for any application. More information available at www.shellab.com
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.