Learn More
Fine Science Tools Silk Suture Thread, Braided Silk, 4/0, 0.17 mm, 23 m
Supplier: Fine Science Tools 1802040
Product: Suture Thread - Braided Silk/Size 4/0/ Size: 4/0/ Suture Diameter: 0.17mm/ Suture Length: 22.5m/ Suture Material: Braided Silk/ Sterile: No/ Unit: Each