Learn More
Med Vet International Oasis PDO Suture Cassette, Size 0, Length of 15M, Each
Supplier: Med Vet International PD10
The veterinary use Oasis PDO suture cassette is a monofilament synthetic absorbable suture. The PDO smooth monofilament suture has a gradual absorption rate between 180 and 220 days. It retains 50% of its tensile strength for up to 6 weeks. This suture provides excellent pliability and holding strength. Size: 0. Length: 15M.