Learn More
Euthanex Surgical Station for Use with EZ Systems S.S. V-shaped Heated Beds and plastic V-shaped non-heated beds.
SDP
Supplier:  Euthanex EZSSVB
Encompass_Preferred
Surgery V-bed station raised platform provides a convenient rest for the wrists during surgery while the patient is stabilized on the bed with sutures. The Surgery Bed Stations raised platform provides a convenient hand rest during surgery while the patient is stabilized on the V-bed with sutures. This stainless steel surgery station allows for optimal lighting, suture tie downs, and adjustable magnifying glass in one compact design. It is compatible with all of the E-Z Anesthesia electrically heated stainless steel and non-heated plastic V-shaped beds. Easy to clean. Features include 304 Stainless steel construction, 3 standard sizes, Non-skid feet, built-in hand rest, LED light with 3 brightness levels, 6X Magnifying glass, built-in suture mounts.
Catalog No. NC2259344