Learn More
Euthanex Surgical Station for Use with EZ Systems S.S. Flat Heated Beds and plastic non-heated beds.
SDP
Supplier:  Euthanex EZSSFB
Encompass_Preferred
Surgery bed station raised platform provides a convenient rest for the wrists during surgery while the patient is stabilized on the bed with sutures. The Surgery Bed Stations raised platform provides a convenient hand rest during surgery while the patient is stabilized on the bed with sutures. This stainless steel surgery station allows for optimal lighting, suture tie downs, and adjustable magnifying glass in one compact design. It is compatible with all of the E-Z Anesthesia electrically heated stainless steel and non-heated plastic beds. Easy to clean. Features include 304 Stainless steel construction, 3 standard sizes, Non-skid feet, built-in hand rest, LED light with 3 brightness levels, 6X Magnifying glass, built-in suture mounts.
Catalog No. NC2259343