Learn More
Surgical Design Safety Scalpels
Top activated retractable safety scalpel
$77.00 - $78.00
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
22-079-716
|
Surgical Design
SSCALPEL#10 |
# 10 |
Pack of 10 for $78.00
|
|
|||||
22-079-718
|
Surgical Design
SSCALPEL#12 |
# 12 |
Pack of 10 for $77.00
|
|
|||||
22-079-719
|
Surgical Design
SSCALPEL#15 |
# 15 |
Pack of 10 for $78.00
|
|
|||||
22-079-720
|
Surgical Design
SSCALPEL#20 |
# 20 |
Pack of 10 for $78.00
|
|
|||||
22-079-717
|
Surgical Design
SSCALPEL#11 |
# 11 |
Pack of 10 for $78.00
|
|
|||||
Description
Top activated retractable safety scalpel
- Audible clicks indicate blade position (retracted, extended and final lock for disposal)
- With non-slip ribbed handle
Specifications
Safety Scalpel |