Learn More
Thermo Scientific™ SureTect™ Cronobacter Species PCR Assay
Rapidly detect Cronobacter species in powdered infant formula and production environment samples in less than 20 hours with minimal handling and maximum confidence in results.
Supplier: Thermo Scientific™ PT1060A
Description
Simply and accurately detect Cronobacter species in enriched powdered infant formula, infant formula raw materials, and production environment samples, in less than 20 hours with the Thermo Scientific™ SureTect™ Cronobacter Species PCR Assay. AFNOR-certified, approved by NF VALIDATION™, and shown to provide negative results two days earlier than the current ISO TS 22964 method, the SureTect Cronobacter Species PCR Assay delivers speed, accuracy and reliability of results. Make faster decisions on product or equipment release while protecting consumers, your brand and your bottom line.
The SureTect System is a unique solution combining speed and performance in an easy-to-use, cost-effective platform—giving you results you can trust.- Rapid actions: fast, optimized single enrichment step and automatic result interpretation.
- Simple workflow: single enrichment protocols, pre-dispensed reagents, universal PCR protocols for detection of multiple targets in the same run.
- Intuitive, user-friendly software: reliable results, with no subjective interpretation.
- Peace of mind: broad range of validated workflows according to AFNOR and AOAC-RI, supported by our dedicated and experienced technical teams
- Maximum flexibility: the ability to run all assays simultaneously and the flexibility to test up to 96 samples in each run helps you operate more efficiently
The Thermo Scientific™ Suretect™ pathogen detection assays are validated to run on the Applied Biosystsmes™ QuantStudio™ 5 Food Safety Real-Time PCR system (Applied Bisosystems™ QuantStudio™ 5 Food Safety Real-Time PCR Instrument with Thermo Scientific™ RapidFinder™ Analysis Software), allowing you to test easily and confidently.
SureTect Cronobacter Species PCR Assay protocols are designed to be simple while producing repeatable, accurate results for production environment samples and dried infant formula (including dried infant formula supplemented with probiotics).
Includes:
- SureTect Lysis Reagent 1 Tubes
- SureTect Lysis Tube Caps
- SureTect Proteinase K
- SureTect Cronobacter species PCR Tubes
- SureTect PCR Caps
Not all products are available for sale in all territories. Please enquire.
Specifications
SureTect Cronobacter Species PCR Assay | |
Powder | |
Fast | |
96 Tests/Kit | |
Real Time PCR (qPCR) |
Primer-probe | |
Low | |
Applied Biosystems 7500 Fast Real-Time PCR instrument and Applied Bisystems QuantStudio 5 Real Time PCR System | |
qPCR Reagents for Microbial Detection |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Laboratory Use Only.