Learn More
Braintree Scientific SureStrain 100m cell strainer, sterile, 1 reducer, yellow
Supplier: Braintree Scientific C40100
SureStrain™ Premium Cell Strainers The SureStrain™ Premium Cell Strainer system is a fast and easy-to-use solution for detaching, isolating and straining cells from clusters and tissues. A polypropylene frame supports a nylon mesh that is available in three color-coded porosities: 40m (blue), 70m (white), and 100m (yellow). The advanced frame/housing design also contributes to superior performance and reduced processing time, by eliminating the air-lock effect. Improved aeration allows air to escape while filtrate is entering the tube, preventing messy spillovers. Advanced design faster flow through Superior ventilation prevents clogging/spills Interlocking and stackable RNase, DNase, pyrogen and cytotoxin free Sterile (gamma) and individually packed Designed for use with 50mL tubes, adapter for 15mL tubes included **Sold for research purposes only and not for human use**
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.