Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
M+A Matting Sure Stride™ Low-Profile Matting
Sure Stride™ is low-profile matting with adhesive backing to ensure it stays in place. Recommended for commercial use on hard floor surfaces to help prevent slips, trips, and falls caused by moisture.
Supplier: M+A Matting 4450013100
Description
- Keeps Floors Dry – Retains up to 1 gallon of water per 10 linear feet (25% more than competitive products)
- Moisture barrier keeps liquids from soaking through to the floor
- Stays in Place – Tacki-Back™ adhesive keeps mat flat and in place, even with heavy cart and buggy traffic
- Safe – Certified high-traction by the National Floor Safety Institute (NFSI)
- Easy to Install – Cut individual mats from the roll as needed
- Dries Quickly – Eco-friendly PET fabric (10 oz.) dissipates moisture and dries quickly
Specifications
Smoke Gray | |
1 Ea. | |
3 ft. |
NFSI Certified. Made in USA | |
3/8 in. | |
100 ft. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction