Learn More
Simport™ Scientific SUPERFLO™ Histology Tissue Cassette, One-Piece Molded, Bulk
This model is a one-piece molded cassette so no separate lids to manage. It has a front opening and a 45° smooth writing surface. It can be opened and closed up to five times guaranteed, securely relocking to avoid specimen loss. Molded from high-density polymer, it keeps specimens safely submerged in liquid.
$472.00
Specifications
Packaging Type | Bulk |
---|---|
Quantity | 3 Boxes of 500 |
Pore Style | 0.91 x 3.67 mm |
Product Type | Tissue Cassette |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
23-045-606
|
Simport™ Scientific
M5302 |
White |
Case of 1500 for $472.00
|
|
|||||
23-045-607
|
Simport™ Scientific
M5303 |
Pink |
Case of 1500 for $472.00
|
|
|||||
23-045-608
|
Simport™ Scientific
M5304 |
Green |
Case of 1500 for $472.00
|
|
|||||
23-045-609
|
Simport™ Scientific
M5305 |
Yellow |
Case of 1500 for $472.00
|
|
|||||
23-045-610
|
Simport™ Scientific
M5306 |
Blue |
Case of 1500 for $472.00
|
|
|||||
23-045-611
|
Simport™ Scientific
M5307 |
Peach |
Case of 1500 for $472.00
|
|
|||||
23-045-612
|
Simport™ Scientific
M5308 |
Tan |
Case of 1500 for $472.00
|
|
|||||
23-045-613
|
Simport™ Scientific
M5309 |
Gray |
Case of 1500 for $472.00
|
|
|||||
23-045-614
|
Simport™ Scientific
M53010 |
Lilac |
Case of 1500 for $472.00
|
|
|||||
23-045-615
|
Simport™ Scientific
M53011 |
Orange |
Case of 1500 for $472.00
|
|
|||||
23-045-616
|
Simport™ Scientific
M53012 |
Aqua |
Case of 1500 for $472.00
|
|
|||||
Description
- Unique lateral vents, fluids can access tissue from all 4 sides
- Basket style for small tissue management
- One-piece molded with front opening
- Smooth 45° angle writing surface
- Available in 11 colors
- This cassette model is supplied in bulk
- IVDR
- CE
Specifications
Bulk | |
3 Boxes of 500 | |
0.91 x 3.67 mm | |
Tissue Cassette |