Learn More
Simport™ Scientific SUPERFLO™ Histology Micro Biopsy Cassette, One-Piece Molded, Bulk
This model is a one-piece molded cassette so no separate lids to manage. It has a front opening and a 45° smooth writing surface. It can be opened and closed up to five times guaranteed, securely relocking to avoid specimen loss. Molded from high-density polymer, it keeps specimens safely submerged in liquid.
$786.00
Specifications
Packaging Type | Bulk |
---|---|
Quantity | 3 Boxes of 500 |
Pore Style | 0.30 x 0.30 mm |
Product Type | Micro Biopsy Cassette |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
23-045-584
|
Simport™ Scientific
M5322 |
White |
Case of 1500 for $786.00
|
|
|||||
23-045-585
|
Simport™ Scientific
M5323 |
Pink |
Case of 1500 for $786.00
|
|
|||||
23-045-586
|
Simport™ Scientific
M5324 |
Green |
Case of 1500 for $786.00
|
|
|||||
23-045-587
|
Simport™ Scientific
M5325 |
Yellow |
Case of 1500 for $786.00
|
|
|||||
23-045-588
|
Simport™ Scientific
M5326 |
Blue |
Case of 1500 for $786.00
|
|
|||||
23-045-589
|
Simport™ Scientific
M5327 |
Peach |
Case of 1500 for $786.00
|
|
|||||
23-045-590
|
Simport™ Scientific
M5328 |
Tan |
Case of 1500 for $786.00
|
|
|||||
23-045-591
|
Simport™ Scientific
M5329 |
Gray |
Case of 1500 for $786.00
|
|
|||||
23-045-592
|
Simport™ Scientific
M53210 |
Lilac |
Case of 1500 for $786.00
|
|
|||||
23-045-593
|
Simport™ Scientific
M53211 |
Orange |
Case of 1500 for $786.00
|
|
|||||
23-045-594
|
Simport™ Scientific
M53212 |
Aqua |
Case of 1500 for $786.00
|
|
|||||
Description
- Unique lateral vents, fluids can access tissue from all 4 sides
- Basket style for small tissue management
- One-piece molded with front opening
- Smooth 45° angle writing surface
- Available in 11 colors
- This cassette model is supplied in bulk
- IVDR
- CE
Specifications
Bulk | |
3 Boxes of 500 | |
0.30 x 0.30 mm | |
Micro Biopsy Cassette |