Learn More
Simport™ Scientific SUPERFLO™ Histology Micro Biopsy Cassette, Hinged Design, Bulk
This model has a special hinge that holds the base and the cover together. The hinge design allows the cassette to be opened and closed as often as required. It is a front-opening cassette with a 45° smooth writing surface. Molded from high-density polymer, keeps specimens safely submerged in liquid.
$786.00
Specifications
Packaging Type | Bulk |
---|---|
Quantity | 3 Boxes of 500 |
Pore Style | 0.30 x 0.30 mm |
Product Type | Micro Biopsy Cassette |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
23-045-683
|
Simport™ Scientific
M5352 |
White |
Case of 1500 for $786.00
|
|
|||||
23-045-684
|
Simport™ Scientific
M5353 |
Pink |
Case of 1500 for $786.00
|
|
|||||
23-045-685
|
Simport™ Scientific
M5354 |
Green |
Case of 1500 for $786.00
|
|
|||||
23-045-686
|
Simport™ Scientific
M5355 |
Yellow |
Case of 1500 for $786.00
|
|
|||||
23-045-687
|
Simport™ Scientific
M5356 |
Blue |
Case of 1500 for $786.00
|
|
|||||
23-045-688
|
Simport™ Scientific
M5357 |
Peach |
Case of 1500 for $786.00
|
|
|||||
23-045-689
|
Simport™ Scientific
M5358 |
Tan |
Case of 1500 for $786.00
|
|
|||||
23-045-690
|
Simport™ Scientific
M5359 |
Gray |
Case of 1500 for $786.00
|
|
|||||
23-045-691
|
Simport™ Scientific
M53510 |
Lilac |
Case of 1500 for $786.00
|
|
|||||
23-045-692
|
Simport™ Scientific
M53511 |
Orange |
Case of 1500 for $786.00
|
|
|||||
23-045-693
|
Simport™ Scientific
M53512 |
Aqua |
Case of 1500 for $786.00
|
|
|||||
Description
- Unique lateral vents, fluids can access tissue from all 4 sides
- Basket style for small tissue management
- Special hinged cover design with front opening
- Smooth 45° angle writing surface
- Available in 11 colors
- This cassette model is supplied in bulk
- IVDR
- CE
Specifications
Bulk | |
3 Boxes of 500 | |
0.30 x 0.30 mm | |
Micro Biopsy Cassette |