Learn More
Simport™ Scientific SUPERFLO™ Histology Biopsy Cassette, One-Piece Molded, Taped
This model is a one-piece molded cassette so no separate lids to manage. It has a front opening and a 45° smooth writing surface. It can be opened and closed up to five times guaranteed, securely relocking to avoid specimen loss. Molded from high-density polymer, it keeps specimens safely submerged in liquid.
$444.00
Specifications
Packaging Type | Taped |
---|---|
Quantity | 25 Stacks of 40 |
Pore Style | 0.64 x 0.64 mm |
Product Type | Biopsy Cassette |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
23-045-716
|
Simport™ Scientific
M4312T |
White |
Case of 1000 for $444.00
|
|
|||||
23-045-717
|
Simport™ Scientific
M4313T |
Pink |
Case of 1000 for $444.00
|
|
|||||
23-045-718
|
Simport™ Scientific
M4314T |
Green |
Case of 1000 for $444.00
|
|
|||||
23-045-719
|
Simport™ Scientific
M4315T |
Yellow |
Case of 1000 for $444.00
|
|
|||||
23-045-720
|
Simport™ Scientific
M4316T |
Blue |
Case of 1000 for $444.00
|
|
|||||
23-045-721
|
Simport™ Scientific
M4317T |
Peach |
Case of 1000 for $444.00
|
|
|||||
23-045-722
|
Simport™ Scientific
M4318T |
Tan |
Case of 1000 for $444.00
|
|
|||||
23-045-723
|
Simport™ Scientific
M4319T |
Gray |
Case of 1000 for $444.00
|
|
|||||
23-045-724
|
Simport™ Scientific
M43110T |
Lilac |
Case of 1000 for $444.00
|
|
|||||
23-045-725
|
Simport™ Scientific
M43111T |
Orange |
Case of 1000 for $444.00
|
|
|||||
23-045-726
|
Simport™ Scientific
M43112T |
Aqua |
Case of 1000 for $444.00
|
|
|||||
Description
- Unique lateral vents, fluids can access tissue from all 4 sides
- Basket style for small tissue management
- One-piece molded with front opening
- Smooth 45° angle writing surface
- Available in 11 colors
- This cassette model is supplied in taped stacks for use with ink jet printers
- IVDR
- CE
Specifications
Taped | |
25 Stacks of 40 | |
0.64 x 0.64 mm | |
Biopsy Cassette |