Learn More
Simport™ Scientific SUPERFLO™ Histology Biopsy Cassette, One-Piece Molded, Bulk
This model is a one-piece molded cassette so no separate lids to manage. It has a front opening and a 45° smooth writing surface. It can be opened and closed up to five times guaranteed, securely relocking to avoid specimen loss. Molded from high-density polymer, it keeps specimens safely submerged in liquid.
$478.00
Specifications
Packaging Type | Bulk |
---|---|
Quantity | 3 Boxes of 500 |
Pore Style | 0.64 x 0.64 mm |
Product Type | Biopsy Cassette |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
23-045-595
|
Simport™ Scientific
M5312 |
White |
Case of 1500 for $478.00
|
|
|||||
23-045-596
|
Simport™ Scientific
M5313 |
Pink |
Case of 1500 for $478.00
|
|
|||||
23-045-597
|
Simport™ Scientific
M5314 |
Green |
Case of 1500 for $478.00
|
|
|||||
23-045-598
|
Simport™ Scientific
M5315 |
Yellow |
Case of 1500 for $478.00
|
|
|||||
23-045-599
|
Simport™ Scientific
M5316 |
Blue |
Case of 1500 for $478.00
|
|
|||||
23-045-600
|
Simport™ Scientific
M5317 |
Peach |
Case of 1500 for $478.00
|
|
|||||
23-045-601
|
Simport™ Scientific
M5318 |
Tan |
Case of 1500 for $478.00
|
|
|||||
23-045-602
|
Simport™ Scientific
M5319 |
Gray |
Case of 1500 for $478.00
|
|
|||||
23-045-603
|
Simport™ Scientific
M53110 |
Lilac |
Case of 1500 for $478.00
|
|
|||||
23-045-604
|
Simport™ Scientific
M53111 |
Orange |
Case of 1500 for $478.00
|
|
|||||
23-045-605
|
Simport™ Scientific
M53112 |
Aqua |
Case of 1500 for $478.00
|
|
|||||
Description
- Unique lateral vents, fluids can access tissue from all 4 sides
- Basket style for small tissue management
- One-piece molded with front opening
- Smooth 45° angle writing surface
- Available in 11 colors
- This cassette model is supplied in bulk
- IVDR
- CE
Specifications
Bulk | |
3 Boxes of 500 | |
0.64 x 0.64 mm | |
Biopsy Cassette |