Learn More
Simport™ Scientific SUPERFLO™ Histology Biopsy Cassette, Hinged Design, Sleeve
This model has a special hinge that holds the base and the cover together. The hinge design allows the cassette to be opened and closed as often as required. It is a front-opening cassette with a 45° smooth writing surface. Molded from high-density polymer, keeps specimens safely submerged in liquid.
$349.00
Specifications
Packaging Type | Sleeve |
---|---|
Quantity | 10 Sleeves of 75 |
Pore Style | 0.64 x 0.64 mm |
Product Type | Biopsy Cassette |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
23-045-661
|
Simport™ Scientific
M5342SL |
White |
Case of 750 for $349.00
|
|
|||||
23-045-662
|
Simport™ Scientific
M5343SL |
Pink |
Case of 750 for $349.00
|
|
|||||
23-045-663
|
Simport™ Scientific
M5344SL |
Green |
Case of 750 for $349.00
|
|
|||||
23-045-664
|
Simport™ Scientific
M5345SL |
Yellow |
Case of 750 for $349.00
|
|
|||||
23-045-665
|
Simport™ Scientific
M5346SL |
Blue |
Case of 750 for $349.00
|
|
|||||
23-045-666
|
Simport™ Scientific
M5347SL |
Peach |
Case of 750 for $349.00
|
|
|||||
23-045-667
|
Simport™ Scientific
M5348SL |
Tan |
Case of 750 for $349.00
|
|
|||||
23-045-668
|
Simport™ Scientific
M5349SL |
Gray |
Case of 750 for $349.00
|
|
|||||
23-045-669
|
Simport™ Scientific
M53410SL |
Lilac |
Case of 750 for $349.00
|
|
|||||
23-045-670
|
Simport™ Scientific
M53411SL |
Orange |
Case of 750 for $349.00
|
|
|||||
23-045-671
|
Simport™ Scientific
M53412SL |
Aqua |
Case of 750 for $349.00
|
|
|||||
Description
- Unique lateral vents, fluids can access tissue from all 4 sides
- Basket style for small tissue management
- Special hinged cover design with front opening
- Smooth 45° angle writing surface
- Available in 11 colors
- This cassette model is supplied in plastic sleeves suitable for printers
- IVDR
- CE
Specifications
Sleeve | |
10 Sleeves of 75 | |
0.64 x 0.64 mm | |
Biopsy Cassette |