Learn More
Simport™ Scientific SUPERFLO™ Histology Biopsy Cassette, Hinged Design, Bulk
This model has a special hinge that holds the base and the cover together. The hinge design allows the cassette to be opened and closed as often as required. It is a front-opening cassette with a 45° smooth writing surface. Molded from high-density polymer, keeps specimens safely submerged in liquid.
$465.00
Specifications
Packaging Type | Bulk |
---|---|
Quantity | 3 Boxes of 500 |
Pore Style | 0.64 x 0.64 mm |
Product Type | Biopsy Cassette |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
23-045-738
|
Simport™ Scientific
M5342 |
White |
Case of 1500 for $465.00
|
|
|||||
23-045-739
|
Simport™ Scientific
M5343 |
Pink |
Case of 1500 for $465.00
|
|
|||||
23-045-740
|
Simport™ Scientific
M5344 |
Green |
Case of 1500 for $465.00
|
|
|||||
23-045-741
|
Simport™ Scientific
M5345 |
Yellow |
Case of 1500 for $465.00
|
|
|||||
23-045-742
|
Simport™ Scientific
M5346 |
Blue |
Case of 1500 for $465.00
|
|
|||||
23-045-743
|
Simport™ Scientific
M5347 |
Peach |
Case of 1500 for $465.00
|
|
|||||
23-045-744
|
Simport™ Scientific
M5348 |
Tan |
Case of 1500 for $465.00
|
|
|||||
23-045-745
|
Simport™ Scientific
M5349 |
Gray |
Case of 1500 for $465.00
|
|
|||||
23-045-746
|
Simport™ Scientific
M53410 |
Lilac |
Case of 1500 for $465.00
|
|
|||||
23-045-747
|
Simport™ Scientific
M53411 |
Orange |
Case of 1500 for $465.00
|
|
|||||
23-045-748
|
Simport™ Scientific
M53412 |
Aqua |
Case of 1500 for $465.00
|
|
|||||
Description
- Unique lateral vents, fluids can access tissue from all 4 sides
- Basket style for small tissue management
- Special hinged cover design with front opening
- Smooth 45° angle writing surface
- Available in 11 colors
- This cassette model is supplied in bulk
- IVDR
- CE
Specifications
Bulk | |
3 Boxes of 500 | |
0.64 x 0.64 mm | |
Biopsy Cassette |