Learn More
WATERBORNE INC SuperStick™ Slides, 2 well, 15mm, 40/pack
Supplier: WATERBORNE INC S1002
Custom-manufactured slides with chemically treated surface to increase adhesion of cysts, oocysts, spores, and other cells. Each slide has a green, Teflon-coated section that is hydrophobic to contain the sample within the well. Each slide also has a super-frosted area at one end for writing with pencil or marker. Packaged forty slides per box.