Learn More
Bioworld Super Optimal Broth (SOB Medium), 100 g
Supplier: Bioworld 306200751
Super Optimal Broth (SOB Medium)Used for cultivation of recombinant strains of E. ColiIs a nutritionally rich growth medium for use in the preparation and transformation of competent cellspH: 7.0Working Concentration: 28 g/LDehydrated Appearance: light beige, homogenous powderPrepared Appearance: light to medium amber agarSoluble in waterContains:Casein Enzymic Hydrolysate Magnesium Sulfate Yeast Extract Potassium Chloride Sodium Chloride
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.