Learn More
Bioworld Super Optimal Broth (SOB Medium), 500 g
Supplier:  Bioworld 306200752
Encompass_Preferred
Super Optimal Broth (SOB Medium)Used for cultivation of recombinant strains of E. ColiIs a nutritionally rich growth medium for use in the preparation and transformation of competent cellspH: 7.0Working Concentration: 28 g/LDehydrated Appearance: light beige, homogenous powderPrepared Appearance: light to medium amber agarSoluble in waterContains:Casein Enzymic Hydrolysate Magnesium Sulfate Yeast Extract Potassium Chloride Sodium Chloride
Catalog No. 50-254-371