Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ SUPELCOSIL™ LC-18 HPLC Column, 5 μm Particle Size
A general purpose hydrophobic alkyl phase that is very retentive and gives good peak shape for a wide variety of compounds.
Supplier: MilliporeSigma™ Supelco™ 58230C40
Description
SUPELCOSIL™ LC-18 HPLC Column may be used for the separation of:. Anthocyanins, flavonols, and carbohydrates in Tropea red onions (Allium cepa L.) during their determination by high performance liquid chromatography (HPLC) as well as HPLC coupled to mass spectrometry (MS) equipped with electrospray ionization (ESI) source, Sinapyl derivatives in pineapple (Ananas comosus [L.] Merill) juice samples during their determination by semipreparative reversed phase HPLC and Aflatoxin M1 in milk and cheese samples during its determination by HPLC with post-column derivatization and fluorescence detection.Specifications
4 mm | |
5 μm | |
170 m2/g | |
SUPELCOSIL | |
Yes |
15 cm | |
120 Å | |
C18 (Octadecyl) | |
C18 (Octadecyl) Phase/Silica Gel, Spherical Particle Platform | |
Reversed Phase |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC
Spot an opportunity for improvement?
Provide Content Correction