Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SUN-SRi™ Step Vial II Unassembled Vial/Cap/Septa Kit

Caps and septa are pre-assembled to save time during sample preparation and to minimize contamination during assembly

Supplier:  Thermo Scientific™ 500 798

Catalog No. 22-030203

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View alternate product


Description

Description

  • Vials are conveniently provided in an adjacent well for easy accessibility Kits are an economical alternative to ordering multiple components
  • 12 x 32mm
  • Maximum fill volume 1.5mL

TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.47 in.
12 mm
1.5 mL
Surestop
32 mm
Yellow
Silica Glass
1.25 in.
Polypropylene
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit