Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 50% v/v (1:1), AquaPhoenix Scientific™
SCI_ED

Catalog Number S88738
Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle,Orange Cap
Poly Bottle,Purple Cap
Poly Bottle,White Cap
7664-93-9, 7732-18-5
H2O4S
98.07
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
OS(O)(=O)=O
7664-93-9, 7732-18-5
H2O4S
98.07
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
OS(O)(=O)=O
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit