Learn More
NSI Environmental Solutions Stable Sulfide Standard 10mg/mL, 6 x 1.5 mL
Supplier: NSI Environmental Solutions QCI147
An exclusive NSI Lab Solutions Product! A sulfide CRM that is stable unopened for 2 years from manufacture. Precipitate free preservation using a proprietary buffer system and inert gas sealing. 10mg/mL Sulfide.