Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sucrose Gel-Loading Dye for DNA Gels, 5X (Contains 40% Sucrose/Mol. Biol.), Fisher BioReagents
Description
This dye includes sucrose instead of glycerol. Add the dye to DNA samples prior to electrophoresis on agarose gels.Specifications
Sucrose Gel-Loading Dye for DNA Gels | |
5X, Contains 40% Sucrose | |
Electrophoresis | |
Purple | |
Liquid |
0.30 to 0.45 AU at 252nm, 0.47 to 0.62 AU, 0.48 to 0.65 AU | |
57-50-1, 7732-18-5 | |
Poly Bottle | |
5 mL |
Safety and Handling
Recommended Storage : RT
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction